missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(tricyclohexylphosphine)nickel(II) Dichloride 95.0+%, TCI America™
$72.70 - $251.65
Chemical Identifiers
| CAS | 19999-87-2 |
|---|---|
| Molecular Formula | C36H66Cl2NiP2 |
| Molecular Weight (g/mol) | 690.46 |
| MDL Number | MFCD11973802 |
| InChI Key | YOCBOYPGZVFUCQ-UHFFFAOYSA-L |
| Synonym | Dichlorobis(tricyclohexylphosphine)nickel(II) |
| PubChem CID | 53384385 |
| IUPAC Name | dichloronickel; bis(tricyclohexylphosphane) |
| SMILES | Cl[Ni]Cl.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1 |
Chemical Identifiers
| 19999-87-2 | |
| 690.46 | |
| YOCBOYPGZVFUCQ-UHFFFAOYSA-L | |
| 53384385 | |
| Cl[Ni]Cl.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1 |
| C36H66Cl2NiP2 | |
| MFCD11973802 | |
| Dichlorobis(tricyclohexylphosphine)nickel(II) | |
| dichloronickel; bis(tricyclohexylphosphane) |
Specifications
| 19999-87-2 | |
| Red-Yellow | |
| MFCD11973802 | |
| Dichlorobis(tricyclohexylphosphine)nickel(II) | |
| Cl[Ni]Cl.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1 | |
| 690.46 | |
| 690.46 | |
| Crystalline Powder |
| 227°C | |
| C36H66Cl2NiP2 | |
| 1 g | |
| YOCBOYPGZVFUCQ-UHFFFAOYSA-L | |
| dichloronickel; bis(tricyclohexylphosphane) | |
| 53384385 | |
| ≥95.0% (T) | |
| Bis(tricyclohexylphosphine)nickel(II) Dichloride |
Safety and Handling
TSCA : No