accessibility menu, dialog, popup

UserName

PPClNiCl

CAS RN 19999-87-2

IUPAC Name: dichloronickel; bis(tricyclohexylphosphane)
Synonyms: nickel chloride; bis(tricyclohexylphosphine)
Molecular Weight (g/mol): 690.46
Molecular Formula: C36H66Cl2NiP2
InChi Key: YOCBOYPGZVFUCQ-UHFFFAOYSA-L
SMILES: Cl[Ni]Cl.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results