missing translation for 'onlineSavingsMsg'
Learn More

Bis(tricyclohexylphosphine)nickel(II) Dichloride 95.0+%, TCI America™

Catalog No. B35341G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35341G 1 g
B35345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35341G Supplier TCI America Supplier No. B35341G

Chemical Identifiers

CAS 19999-87-2
Molecular Formula C36H66Cl2NiP2
Molecular Weight (g/mol) 690.46
MDL Number MFCD11973802
InChI Key YOCBOYPGZVFUCQ-UHFFFAOYSA-L
Synonym Dichlorobis(tricyclohexylphosphine)nickel(II)
PubChem CID 53384385
IUPAC Name dichloronickel; bis(tricyclohexylphosphane)
SMILES Cl[Ni]Cl.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1

Specifications

Melting Point 227°C
Color Red-Yellow
Quantity 1 g
Formula Weight 690.46
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(tricyclohexylphosphine)nickel(II) Dichloride