missing translation for 'onlineSavingsMsg'
Learn More

Bis(tri-tert-butylphosphine)palladium(0) 98.0+%, TCI America™

Catalog No. p-7119992
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3161250MG 250 mg
B31611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3161250MG Supplier TCI America Supplier No. B3161250MG
Only null left

Chemical Identifiers

CAS 53199-31-8
Molecular Formula C24H54P2Pd
Molecular Weight (g/mol) 511.06
MDL Number MFCD03094580
InChI Key MXQOYLRVSVOCQT-UHFFFAOYSA-N
Synonym bis tri-tert-butylphosphine palladium 0, bis tri-t-butylphosphine palladium 0, bis tri-tert-butylphosphine palladium, bis tri-tert-butylphosphane palladium, palladium; tritert-butylphosphane, palladium, bis tris 1,1-dimethylethyl phosphine, pd p tbu 3 2, pd t-bu3p 2, bis tri-t-butylphosphine palladium, di tri-tert-butylphosphine palladium 0
PubChem CID 2734558
IUPAC Name bis(tri-tert-butylphosphane) palladium
SMILES [Pd].CC(C)(C)P(C(C)(C)C)C(C)(C)C.CC(C)(C)P(C(C)(C)C)C(C)(C)C

Specifications

Quantity 250 mg
Formula Weight 511.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(tri-tert-butylphosphine)palladium(0)
TSCA No
Recommended Storage Freezer