Learn More
Bis(tri-tert-butylphosphine)palladium(0), 98%
CAS: 53199-31-8 | C24H54P2Pd | 511.06 g/mol
$588.98 - $588.98
Chemical Identifiers
| CAS | 53199-31-8 |
|---|---|
| Molecular Formula | C24H54P2Pd |
| Molecular Weight (g/mol) | 511.06 |
| MDL Number | MFCD03094580 |
| InChI Key | MXQOYLRVSVOCQT-UHFFFAOYSA-N |
| Synonym | bis tri-tert-butylphosphine palladium 0, bis tri-t-butylphosphine palladium 0, bis tri-tert-butylphosphine palladium, bis tri-tert-butylphosphane palladium, palladium; tritert-butylphosphane, palladium, bis tris 1,1-dimethylethyl phosphine, pd p tbu 3 2, pd t-bu3p 2, bis tri-t-butylphosphine palladium, di tri-tert-butylphosphine palladium 0 |
| PubChem CID | 2734558 |
| SMILES | [Pd].CC(C)(C)P(C(C)(C)C)C(C)(C)C.CC(C)(C)P(C(C)(C)C)C(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC363500010
|
Thermo Scientific Chemicals
363500010 |
1 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 53199-31-8 | |
| 511.06 | |
| MXQOYLRVSVOCQT-UHFFFAOYSA-N | |
| 2734558 |
| C24H54P2Pd | |
| MFCD03094580 | |
| bis tri-tert-butylphosphine palladium 0, bis tri-t-butylphosphine palladium 0, bis tri-tert-butylphosphine palladium, bis tri-tert-butylphosphane palladium, palladium; tritert-butylphosphane, palladium, bis tris 1,1-dimethylethyl phosphine, pd p tbu 3 2, pd t-bu3p 2, bis tri-t-butylphosphine palladium, di tri-tert-butylphosphine palladium 0 | |
| [Pd].CC(C)(C)P(C(C)(C)C)C(C)(C)C.CC(C)(C)P(C(C)(C)C)C(C)(C)C |
Specifications
| 53199-31-8 | |
| Brown-Orange to White | |
| 97.5% min. (ex Pd) | |
| C24H54P2Pd | |
| 1 g | |
| Solubility in water: insoluble | |
| [Pd].CC(C)(C)P(C(C)(C)C)C(C)(C)C.CC(C)(C)P(C(C)(C)C)C(C)(C)C | |
| 511.06 | |
| 511.05 | |
| Crystals |
| 258.0°C to 272.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD03094580 | |
| bis tri-tert-butylphosphine palladium 0, bis tri-t-butylphosphine palladium 0, bis tri-tert-butylphosphine palladium, bis tri-tert-butylphosphane palladium, palladium; tritert-butylphosphane, palladium, bis tris 1,1-dimethylethyl phosphine, pd p tbu 3 2, pd t-bu3p 2, bis tri-t-butylphosphine palladium, di tri-tert-butylphosphine palladium 0 | |
| MXQOYLRVSVOCQT-UHFFFAOYSA-N | |
| palladium;tritert-butylphosphane | |
| 2734558 | |
| 98% | |
| Bis(tri-tert-butylphosphine)palladium(0) |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
RUO – Research Use Only