missing translation for 'onlineSavingsMsg'
Learn More

Bis(trans-3,3,5-trimethylcyclohexyl) Phthalate 95.0+%, TCI America™

Catalog No. p069725g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P069725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P069725G Supplier TCI America Supplier No. P069725G

Chemical Identifiers

CAS 245652-82-8
Molecular Formula C26H38O4
Molecular Weight (g/mol) 414.586
MDL Number MFCD00070481
InChI Key ATHBXDPWCKSOLE-ADEKHHIBSA-N
Synonym Phthalic Acid Bis(trans-3,3,5-trimethylcyclohexyl) Ester
PubChem CID 91188092
IUPAC Name 2-O-[(1R,5S)-3,3,5-trimethylcyclohexyl] 1-O-(3,3,5-trimethylcyclohexyl) benzene-1,2-dicarboxylate
SMILES CC1CC(CC(C1)(C)C)OC(=O)C2=CC=CC=C2C(=O)OC3CC(CC(C3)(C)C)C

Specifications

Melting Point 54°C
Color White
Quantity 25 g
Formula Weight 414.59
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(trans-3,3,5-trimethylcyclohexyl) Phthalate