missing translation for 'onlineSavingsMsg'
Learn More

Bis(tetraethylammonium) bis(2-thioxo-1,3-dithiole-4,5-dithiolato)zincate, 97%

Catalog No. p-7046987
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL17046MD 250 mg
AAL1704603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL17046MD Supplier Thermo Scientific Chemicals Supplier No. L17046MD
Only null left

CAS: 72022-68-5 | C22H40N2S10Zn | 718.556 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 72022-68-5
Molecular Formula C22H40N2S10Zn
Molecular Weight (g/mol) 718.556
MDL Number MFCD00623815
InChI Key TXRIVAQWJHZFOT-UHFFFAOYSA-J
Synonym bis tetraethylammonium bis 2-thioxo-1,3-dithiole-4,5-dithiolato zincate
PubChem CID 15762389
IUPAC Name zinc;2-sulfanylidene-1,3-dithiole-4,5-dithiolate;tetraethylazanium
SMILES CC[N+](CC)(CC)CC.CC[N+](CC)(CC)CC.C1(=C(SC(=S)S1)[S-])[S-].C1(=C(SC(=S)S1)[S-])[S-].[Zn+2]

Specifications

Melting Point 202°C to 206°C
Quantity 250 mg
Beilstein 4900651
Sensitivity Light sensitive
Formula Weight 718.6
Percent Purity 97%
Chemical Name or Material Bis(tetraethylammonium) bis(2-thioxo-1,3-dithiole-4,5-dithiolato)zincate
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only