missing translation for 'onlineSavingsMsg'
Learn More

Bis(tetrabutylammonium) Bis(1,3-dithiole-2-thione-4,5-dithiolato)palladium(II) 90.0+%, TCI America™

Catalog No. B1437100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1437100MG 100 mg
B14371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1437100MG Supplier TCI America Supplier No. B1437100MG

Chemical Identifiers

CAS 72688-90-5
Molecular Formula C38H72N2PdS10
Molecular Weight (g/mol) 984.028
MDL Number MFCD00143779
InChI Key GKUMQKGIGICMGT-UHFFFAOYSA-J
PubChem CID 15447064
IUPAC Name palladium(2+);2-sulfanylidene-1,3-dithiole-4,5-dithiolate;tetrabutylazanium
SMILES CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.C1(=C(SC(=S)S1)[S-])[S-].C1(=C(SC(=S)S1)[S-])[S-].[Pd+2]

Specifications

Melting Point 205°C
Color Black-Purple
Quantity 100 mg
Formula Weight 984.03
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(tetrabutylammonium) Bis(1,3-dithiole-2-thione-4,5-dithiolato)palladium(II)