accessibility menu, dialog, popup

UserName

H3CNCH3H3CH3CH3CNCH3H3CH3CSSSSSSSSSSPd

CAS RN 72688-90-5

IUPAC Name: palladium(2+) bis((5-sulfanidyl-2-sulfanylidene-2H-1,3-dithiol-4-yl)sulfanide) bis(tetrabutylazanium)
Synonyms: palladium(2+) bis((5-sulfanidyl-2-sulfanylidene-1,3-dithiol-4-yl)sulfanide) bis(tetrabutylammonium ion)
Molecular Weight (g/mol): 984.03
Molecular Formula: C38H72N2PdS10
InChi Key: GKUMQKGIGICMGT-UHFFFAOYSA-J
SMILES: [Pd++].[S-]C1=C([S-])SC(=S)S1.[S-]C1=C([S-])SC(=S)S1.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results