missing translation for 'onlineSavingsMsg'
Learn More

(-)-Bis[(S)-1-phenylethyl]amine, ChiPros™, 99%, ee 98+%

Catalog No. AAH2710603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2710603 1 g
AAH2710606 5 g
AAH2710614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2710603 Supplier Thermo Scientific Chemicals Supplier No. H2710603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 56210-72-1 | C16H19N | 225.335 g/mol

(-)-Bis[(S)-1-phenylethyl]amine is used as a chiral resolution reagent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-Bis[(S)-1-phenylethyl]amine is used as a chiral resolution reagent in organic synthesis.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 56210-72-1
Molecular Formula C16H19N
Molecular Weight (g/mol) 225.335
MDL Number MFCD00243087
InChI Key NXLACVVNHYIYJN-KBPBESRZSA-N
Synonym --bis s-1-phenylethyl amine, s-bis s-1-phenylethyl amine, unii-5d97t2y7wu, bis s-1-phenylethyl amine, 1s-1-phenyl-n-1s-1-phenylethyl ethanamine, bis 1s-1-phenylethyl amine, s,s-di 1-phenylethyl amine
PubChem CID 6994958
IUPAC Name (1S)-1-phenyl-N-[(1S)-1-phenylethyl]ethanamine
SMILES CC(C1=CC=CC=C1)NC(C)C2=CC=CC=C2

Specifications

Boiling Point 98°C (0.3 mmHg)
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Immiscible with water.
Formula Weight 225.34
Percent Purity 99%
Assay ee ≥98%
Chemical Name or Material (-)-Bis[(S)-1-phenylethyl]amine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only