accessibility menu, dialog, popup

UserName

CAS RN 56210-72-1

IUPAC Name: bis(1-phenylethyl)amine
Synonyms: bis(α-methylbenzyl)amine
Molecular Weight (g/mol): 225.34
Molecular Formula: C16H19N
InChi Key: NXLACVVNHYIYJN-UHFFFAOYNA-N
SMILES: CC(NC(C)C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)