missing translation for 'onlineSavingsMsg'
Learn More

Bis(phenylacetyl) disulfide, 98%

Catalog No. p-7047811
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1979206 5 g
AAL1979214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1979206 Supplier Thermo Scientific Chemicals Supplier No. L1979206
Only null left

CAS: 15088-78-5 | C16H14O2S2 | 302.41 g/mol

Bis(phenylacetyl)disulfide as sulfur substitution agent in preparation of RNA oligonucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(phenylacetyl)disulfide as sulfur substitution agent in preparation of RNA oligonucleotides.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 15088-78-5
Molecular Formula C16H14O2S2
Molecular Weight (g/mol) 302.41
MDL Number MFCD00513572
InChI Key IXGZXXBJSZISOO-UHFFFAOYSA-N
Synonym phenylacetyl disulfide, 2-phenylacetic dithioperoxyanhydride, phenylacetyl disulphide, bis phenylacetyl disulfide, bis phenylacetyl disulphide, 2-phenyl-1-2-phenylacetyl disulfanyl ethanone, bis phenylacetyl persulfide, phenylacetyl disulfide 5g, s-2-phenylacetyl sulfanyl 2-phenylethanethioate
PubChem CID 3378603
IUPAC Name S-(2-phenylacetyl)sulfanyl 2-phenylethanethioate
SMILES O=C(CC1=CC=CC=C1)SSC(=O)CC1=CC=CC=C1

Specifications

Melting Point 58°C to 60°C
Quantity 5 g
Beilstein 2057579
Solubility Information Insoluble in water.
Formula Weight 302.42
Percent Purity 98%
Chemical Name or Material Bis(phenylacetyl) disulfide
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only