missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(pentamethylcyclopentadienyl)nickel
CAS: 74507-63-4 | C20H30Ni | 329.153 g/mol
$405.64 - $1528.56
Chemical Identifiers
| CAS | 74507-63-4 |
|---|---|
| Molecular Formula | C20H30Ni |
| Molecular Weight (g/mol) | 329.153 |
| MDL Number | MFCD00058704 |
| InChI Key | CHPLEWYRKUFKQP-UHFFFAOYSA-N |
| Synonym | Decamethylnickelocene |
| PubChem CID | 24943158 |
| IUPAC Name | nickel(2+);1,2,3,5,5-pentamethylcyclopenta-1,3-diene |
| SMILES | CC1=[C-]C(C(=C1C)C)(C)C.CC1=[C-]C(C(=C1C)C)(C)C.[Ni+2] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 74507-63-4 | |
| 329.153 | |
| CHPLEWYRKUFKQP-UHFFFAOYSA-N | |
| 24943158 | |
| CC1=[C-]C(C(=C1C)C)(C)C.CC1=[C-]C(C(=C1C)C)(C)C.[Ni+2] |
| C20H30Ni | |
| MFCD00058704 | |
| Decamethylnickelocene | |
| nickel(2+);1,2,3,5,5-pentamethylcyclopenta-1,3-diene |
Specifications
| 74507-63-4 | |
| C20H30Ni | |
| 1 g | |
| Decamethylnickelocene | |
| CC1=[C-]C(C(=C1C)C)(C)C.CC1=[C-]C(C(=C1C)C)(C)C.[Ni+2] | |
| 329.153 | |
| 329.17 | |
| Bis(pentamethylcyclopentadienyl)nickel |
| Ampule | |
| MFCD00058704 | |
| Air and moisture sensitive | |
| CHPLEWYRKUFKQP-UHFFFAOYSA-N | |
| nickel(2+);1,2,3,5,5-pentamethylcyclopenta-1,3-diene | |
| 24943158 | |
| Crystalline |
Safety and Handling
GHS H Statement
H350
May cause cancer.
P201-P202-P281-P308+P313-P501c
H350
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only