accessibility menu, dialog, popup

UserName

CH3CCH3H3CNiCCH3CCH3CCH3CH3CCCH3H3CH3C

CAS RN 74507-63-4

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 329.16
Molecular Formula: C20H30Ni
InChi Key: JLYFAOFYDLSGOE-UHFFFAOYSA-N
SMILES: Cc1c(C)c(C)[c-](C)c1C.C[C-]12[C-]3(C)[C-]4(C)[C-]5(C)[C-]1(C)[Ni]2345
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results