missing translation for 'onlineSavingsMsg'
Learn More

Bis(pentamethylcyclopentadienyl)nickel

Catalog No. AA3923806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3923806 5 g
AA3923803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3923806 Supplier Thermo Scientific Chemicals Supplier No. 03923806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 74507-63-4 | C20H30Ni | 329.153 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 74507-63-4
Molecular Formula C20H30Ni
Molecular Weight (g/mol) 329.153
MDL Number MFCD00058704
InChI Key CHPLEWYRKUFKQP-UHFFFAOYSA-N
Synonym Decamethylnickelocene
PubChem CID 24943158
IUPAC Name nickel(2+);1,2,3,5,5-pentamethylcyclopenta-1,3-diene
SMILES CC1=[C-]C(C(=C1C)C)(C)C.CC1=[C-]C(C(=C1C)C)(C)C.[Ni+2]

Specifications

Packaging Ampule
Quantity 5 g
Sensitivity Air and moisture sensitive
Formula Weight 329.17
Physical Form Crystalline
Chemical Name or Material Bis(pentamethylcyclopentadienyl)nickel
Hazard Category H350
Hazard Statement GHS H Statement
H350
May cause cancer.
Precautionary Statement P201-P202-P281-P308+P313-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only