missing translation for 'onlineSavingsMsg'
Learn More

Bis(pentafluorophenyl)phenylphosphine, 90%, tech., Thermo Scientific™

Catalog No. AC313171000
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC313171000 100 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC313171000 Supplier Thermo Scientific Chemicals Supplier No. 313171000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5074-71-5
Molecular Formula C18H5F10P
Molecular Weight (g/mol) 442.20
InChI Key OYNXPGGNQMSMTR-UHFFFAOYSA-N
Synonym bis pentafluorophenyl phenylphosphine, dftpp, decafluorotriphenylphosphine, bis perfluorophenyl phenyl phosphine, bis 2,3,4,5,6-pentafluorophenyl-phenylphosphane, phosphine,bis 2,3,4,5,6-pentafluorophenyl phenyl, bis 2,3,4,5,6-pentafluorophenyl phenyl phosphane, ultramark 443, bis pentafluorophenyl phenyl phosphite, bis pentafluorophenyl phenyl phosphine
PubChem CID 78757
IUPAC Name bis(2,3,4,5,6-pentafluorophenyl)-phenylphosphane
SMILES FC1=C(F)C(F)=C(P(C2=CC=CC=C2)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F

Specifications

Melting Point 56.0°C to 62.0°C
Boiling Point 105.0°C (0.2 mmHg)
Flash Point >110°C
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula (C6F5)2PC6H5
Quantity 100 mg
Solubility Information Solubility in water: insoluble in water.
Formula Weight 442.2
Percent Purity 90%
Chemical Name or Material Bis(pentafluorophenyl)phenylphosphine
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 225-780-1

RUO – Research Use Only