missing translation for 'onlineSavingsMsg'
Learn More

Bis(neopentyl Glycolato)diboron 98.0+%, TCI America™

Catalog No. p-7119581
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22541G 1 g
B22545G 5 g
B225425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B22541G Supplier TCI America Supplier No. B22541G
Only null left

Chemical Identifiers

CAS 201733-56-4
Molecular Formula C10H20B2O4
Molecular Weight (g/mol) 225.89
MDL Number MFCD02093062
InChI Key MDNDJMCSXOXBFZ-UHFFFAOYSA-N
Synonym bis neopentyl glycolato diboron, bis neopentylglycolato diboron, 5,5,5',5'-tetramethyl-2,2'-bi 1,3,2-dioxaborinane, 2-5,5-dimethyl-1,3,2-dioxaborinan-2-yl-5,5-dimethyl-1,3,2-dioxaborinane, 5,5,5',5'-tetramethyl-2,2'-bi-1,3,2-dioxaborinane, 2,2'-bi-1,3,2-dioxaborinane, 5,5,5',5'-tetramethyl, bis neopentyl glycolato diborane, bis 2,2-dimethyl-1,3-propanediolato diboron, 5,5,5',5'-tetramethyl-2,2' bi 1,3,2 dioxaborinanyl
PubChem CID 2734316
IUPAC Name 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,5-dimethyl-1,3,2-dioxaborinane
SMILES CC1(C)COB(OC1)B1OCC(C)(C)CO1

Specifications

Melting Point 192°C
Color White-Yellow
Quantity 1 g
Formula Weight 225.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(neopentyl Glycolato)diboron
TSCA No
Recommended Storage Refrigerator