accessibility menu, dialog, popup

UserName

CH3H3COBOBOCH3H3CO

CAS RN 201733-56-4

IUPAC Name: 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,5-dimethyl-1,3,2-dioxaborinane
Synonyms: 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,5-dimethyl-1,3,2-dioxaborinane
Molecular Weight (g/mol): 225.89
Molecular Formula: C10H20B2O4
InChi Key: MDNDJMCSXOXBFZ-UHFFFAOYSA-N
SMILES: CC1(C)COB(OC1)B1OCC(C)(C)CO1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)