missing translation for 'onlineSavingsMsg'
Learn More

Bis(methylthio)methane 98.0+%, TCI America™

Catalog No. B12045ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12045ML 5 mL
B120425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12045ML Supplier TCI America Supplier No. B12045ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1618-26-4
Molecular Formula C3H8S2
Molecular Weight (g/mol) 108.217
MDL Number MFCD00008564
InChI Key LOCDPORVFVOGCR-UHFFFAOYSA-N
Synonym bis methylthio methane, bis methylsulfanyl methane, 2,4-dithiapentane, methane, bis methylthio, bis methylmercapto methane, formaldehyde dimethyl mercaptal, methylenebis methyl sulfide, thioformaldehyde dimethylacetal, ch3sch2sch3
PubChem CID 15380
IUPAC Name bis(methylsulfanyl)methane
SMILES CSCSC

Specifications

Boiling Point 152°C
Quantity 5 mL
UN Number 1993
Formula Weight 108.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(methylthio)methane