missing translation for 'onlineSavingsMsg'
Learn More

Bis(methylthio)methane, 99%

CAS: 1618-26-4 | C3H8S2 | 108.217 g/mol

$160.12 - $160.12

Chemical Identifiers

CAS 1618-26-4
Molecular Formula C3H8S2
Molecular Weight (g/mol) 108.217
MDL Number MFCD00008564
InChI Key LOCDPORVFVOGCR-UHFFFAOYSA-N
Synonym bis methylthio methane, bis methylsulfanyl methane, 2,4-dithiapentane, methane, bis methylthio, bis methylmercapto methane, formaldehyde dimethyl mercaptal, methylenebis methyl sulfide, thioformaldehyde dimethylacetal, ch3sch2sch3
PubChem CID 15380
IUPAC Name bis(methylsulfanyl)methane
SMILES CSCSC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1970614
View Documents
Thermo Scientific Chemicals
A1970614
25 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1618-26-4
108.217
LOCDPORVFVOGCR-UHFFFAOYSA-N
15380
CSCSC
C3H8S2
MFCD00008564
bis methylthio methane, bis methylsulfanyl methane, 2,4-dithiapentane, methane, bis methylthio, bis methylmercapto methane, formaldehyde dimethyl mercaptal, methylenebis methyl sulfide, thioformaldehyde dimethylacetal, ch3sch2sch3
bis(methylsulfanyl)methane

Specifications

1618-26-4
146°C to 147°C
Unpleasant
1.534
25 g
1731143
bis methylthio methane, bis methylsulfanyl methane, 2,4-dithiapentane, methane, bis methylthio, bis methylmercapto methane, formaldehyde dimethyl mercaptal, methylenebis methyl sulfide, thioformaldehyde dimethylacetal, ch3sch2sch3
CSCSC
108.217
108.23
Bis(methylthio)methane
1.059
43°C (109°F)
C3H8S2
MFCD00008564
UN1993
14,1256
LOCDPORVFVOGCR-UHFFFAOYSA-N
bis(methylsulfanyl)methane
15380
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 216-577-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only