missing translation for 'onlineSavingsMsg'
Learn More

Bis(hexamethylene)triamine 95.0+%, TCI America™

Catalog No. b1814400g
Change view
Click to view available options
Quantity:
25 g
400 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1814400G 400 g
B181425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1814400G Supplier TCI America Supplier No. B1814400G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 143-23-7
Molecular Formula C12H29N3
Molecular Weight (g/mol) 215.39
MDL Number MFCD00010448
InChI Key MRNZSTMRDWRNNR-UHFFFAOYSA-N
Synonym bis hexamethylene triamine, bis 6-aminohexyl amine, dihexylenetriamine, 1,13-diamino-7-azatridecane, 7-azatridecane-1,13-diamine, 6,6'-diaminodihexylamine, n-6-aminohexyl hexane-1,6-diamine, 1,6-hexanediamine, n-6-aminohexyl, bishexamethylenetriamine, unii-bid902uf5v
PubChem CID 8924
IUPAC Name bis(6-aminohexyl)amine
SMILES NCCCCCCNCCCCCCN

Specifications

Melting Point 35°C
Color White-Yellow
Boiling Point 164°C
Quantity 400 g
UN Number 3259
Formula Weight 215.39
Percent Purity ≥95.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Bis(hexamethylene)triamine
EINECSNumber (2)-2376
RTECSNumber MO1186250
TSCA Yes