accessibility menu, dialog, popup

UserName

H2NNHNH2

CAS RN 143-23-7

IUPAC Name: bis(6-aminohexyl)amine
Synonyms: bis(hexamethylene)triamine
Molecular Weight (g/mol): 215.39
Molecular Formula: C12H29N3
InChi Key: MRNZSTMRDWRNNR-UHFFFAOYSA-N
SMILES: NCCCCCCNCCCCCCN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results