missing translation for 'onlineSavingsMsg'
Learn More

Bis(hexafluoroacetylacetonato)copper(II) Hydrate 95.0+%, TCI America™

Catalog No. H05541G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05541G 1 g
H05545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05541G Supplier TCI America Supplier No. H05541G

Chemical Identifiers

CAS 14781-45-4
Molecular Formula C10H4CuF12O5
Molecular Weight (g/mol) 495.664
MDL Number MFCD00064754
InChI Key SREVICRJIZZXHD-SUXDNRKISA-L
Synonym Copper(II) Hexafluoroacetylacetonate, Hexafluoroacetylacetono Copper(II) Salt
PubChem CID 15860928
IUPAC Name copper;(Z)-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate;hydrate
SMILES C(=C(C(F)(F)F)[O-])C(=O)C(F)(F)F.C(=C(C(F)(F)F)[O-])C(=O)C(F)(F)F.O.[Cu+2]

Specifications

Color Green
Quantity 1 g
Formula Weight 477.65
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(hexafluoroacetylacetonato)copper(II) Hydrate