missing translation for 'onlineSavingsMsg'
Learn More

Bis(hexafluoroacetylacetonato)cobalt(II) Hydrate 95.0+%, TCI America™

Catalog No. H05535G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H05535G Supplier TCI America Supplier No. H05535G
Only null left

Chemical Identifiers

CAS 19648-83-0
Molecular Formula C10H6CoF12O5
Molecular Weight (g/mol) 493.07
MDL Number MFCD00150649,MFCD00042510
InChI Key POHWVOSIFCILEF-UHFFFAOYSA-N
Synonym bis hexafluoroacetylacetonato cobalt ii hydrate, cobalt 2+ bis 1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate, cobalt 2+ ; z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate
PubChem CID 16212172
IUPAC Name bis(1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one) cobalt hydrate
SMILES O.[Co].OC(=CC(=O)C(F)(F)F)C(F)(F)F.OC(=CC(=O)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 197°C
Quantity 5 g
Formula Weight 473.04
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(hexafluoroacetylacetonato)cobalt(II) Hydrate