accessibility menu, dialog, popup

UserName

CAS RN 19648-83-0

FFFHOOFFFFFFHOOFFFH2OCo(E/Z)(E/Z)

CAS RN 19648-83-0

IUPAC Name: bis(1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one) cobalt hydrate
Synonyms: bis(1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one) cobalt hydrate
Molecular Weight (g/mol): 493.07
Molecular Formula: C10H6CoF12O5
InChi Key: POHWVOSIFCILEF-UHFFFAOYSA-N
SMILES: O.[Co].OC(=CC(=O)C(F)(F)F)C(F)(F)F.OC(=CC(=O)C(F)(F)F)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results