missing translation for 'onlineSavingsMsg'
Learn More

Bis(diphenylphosphino)methane 97.0+%, TCI America™

Catalog No. p-7119399
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19825G 5 g
B198225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19825G Supplier TCI America Supplier No. B19825G
Only null left

Chemical Identifiers

CAS 2071-20-7
Molecular Formula C25H22P2
Molecular Weight (g/mol) 384.40
MDL Number MFCD00003537
InChI Key XGCDBGRZEKYHNV-UHFFFAOYSA-N
Synonym bis diphenylphosphino methane, dppm, methylenebis diphenylphosphine, phosphine, methylenebis diphenyl, tetraphenylmethylenediphosphine, unii-i5o2hzr38v, methanediylbis diphenylphosphine, diphenylphosphanyl methyl diphenylphosphane, 1,1-bis diphenylphosphino methane
PubChem CID 74952
IUPAC Name [(diphenylphosphanyl)methyl]diphenylphosphane
SMILES C(P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 120°C
Color White-Yellow
Quantity 5 g
Formula Weight 384.40
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(diphenylphosphino)methane
EINECSNumber (3)-3972
TSCA No