accessibility menu, dialog, popup

UserName

PP

CAS RN 2071-20-7

IUPAC Name: [(diphenylphosphanyl)methyl]diphenylphosphane
Synonyms: bis(diphenylphosphino)methane
Molecular Weight (g/mol): 384.4
Molecular Formula: C25H22P2
InChi Key: XGCDBGRZEKYHNV-UHFFFAOYSA-N
SMILES: C(P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)