Learn More
Bis(diphenylphosphino)methane, 97%
CAS: 2071-20-7 | C25H22P2 | 384.40 g/mol
$744.96 - $744.96
Chemical Identifiers
| CAS | 2071-20-7 |
|---|---|
| Molecular Formula | C25H22P2 |
| Molecular Weight (g/mol) | 384.40 |
| MDL Number | MFCD00003537 |
| InChI Key | XGCDBGRZEKYHNV-UHFFFAOYSA-N |
| Synonym | bis diphenylphosphino methane, dppm, methylenebis diphenylphosphine, phosphine, methylenebis diphenyl, tetraphenylmethylenediphosphine, unii-i5o2hzr38v, methanediylbis diphenylphosphine, diphenylphosphanyl methyl diphenylphosphane, 1,1-bis diphenylphosphino methane |
| PubChem CID | 74952 |
| IUPAC Name | diphenylphosphanylmethyl(diphenyl)phosphane |
| SMILES | C(P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2111218
|
Thermo Scientific Chemicals
B2111218 |
50 g |
Each for $744.96
|
|
|||||
Description
It is applied as an organic synthesis catalyst. It is also used as an intermediate in flavor and fragrances.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is applied as an organic synthesis catalyst. It is also used as an intermediate in flavor and fragrances.
Solubility
Insoluble in water.
Notes
Air sensitive. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 2071-20-7 | |
| 384.40 | |
| XGCDBGRZEKYHNV-UHFFFAOYSA-N | |
| 74952 | |
| C(P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
| C25H22P2 | |
| MFCD00003537 | |
| bis diphenylphosphino methane, dppm, methylenebis diphenylphosphine, phosphine, methylenebis diphenyl, tetraphenylmethylenediphosphine, unii-i5o2hzr38v, methanediylbis diphenylphosphine, diphenylphosphanyl methyl diphenylphosphane, 1,1-bis diphenylphosphino methane | |
| diphenylphosphanylmethyl(diphenyl)phosphane |
Specifications
| 2071-20-7 | |
| C25H22P2 | |
| 50 g | |
| Air Sensitive | |
| Insoluble in water. | |
| C(P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 384.40 | |
| 384.4 | |
| Bis(diphenylphosphino)methane |
| 118°C to 119°C | |
| MFCD00003537 | |
| 758760 | |
| bis diphenylphosphino methane, dppm, methylenebis diphenylphosphine, phosphine, methylenebis diphenyl, tetraphenylmethylenediphosphine, unii-i5o2hzr38v, methanediylbis diphenylphosphine, diphenylphosphanyl methyl diphenylphosphane, 1,1-bis diphenylphosphino methane | |
| XGCDBGRZEKYHNV-UHFFFAOYSA-N | |
| diphenylphosphanylmethyl(diphenyl)phosphane | |
| 74952 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 218-194-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only