missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(dimethylamino)dimethylsilane 97.0+%, TCI America™
Supplier: TCI America B215025ML
Specifications
| Bis(dimethylamino)dimethylsilane | |
| Yellow | |
| C6H18N2Si | |
| 25 mL | |
| bis dimethylamino dimethylsilane, silanediamine, hexamethyl, silanediamine, n,n,n',n',1,1-hexamethyl, acmc-1ctl2, bis-dimethylamino dimethylsilane, n,n,n,n-hexamethylsilanediamine #, dimethylsilane, bis n,n-dimethylamino, n,n,n',n',1,1-hexamethylsilanediamine | |
| CN(C)[Si](C)(C)N(C)C | |
| 146.309 | |
| 146.31 | |
| Liquid |
| 3768-58-9 | |
| 128°C | |
| MFCD00008296 | |
| 2924 | |
| QULMGWCCKILBTO-UHFFFAOYSA-N | |
| N-[dimethylamino(dimethyl)silyl]-N-methylmethanamine | |
| 77384 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 3768-58-9 | |
| 146.309 | |
| QULMGWCCKILBTO-UHFFFAOYSA-N | |
| 77384 | |
| CN(C)[Si](C)(C)N(C)C |
| C6H18N2Si | |
| MFCD00008296 | |
| bis dimethylamino dimethylsilane, silanediamine, hexamethyl, silanediamine, n,n,n',n',1,1-hexamethyl, acmc-1ctl2, bis-dimethylamino dimethylsilane, n,n,n,n-hexamethylsilanediamine #, dimethylsilane, bis n,n-dimethylamino, n,n,n',n',1,1-hexamethylsilanediamine | |
| N-[dimethylamino(dimethyl)silyl]-N-methylmethanamine |
Safety and Handling
TSCA : Yes