missing translation for 'onlineSavingsMsg'
Learn More

Bis(dimethylamino)dimethylsilane 97.0+%, TCI America™

Catalog No. p-7119513
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B215025ML 25 mL
B2150250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B215025ML Supplier TCI America Supplier No. B215025ML
Only null left

Chemical Identifiers

CAS 3768-58-9
Molecular Formula C6H18N2Si
Molecular Weight (g/mol) 146.309
MDL Number MFCD00008296
InChI Key QULMGWCCKILBTO-UHFFFAOYSA-N
Synonym bis dimethylamino dimethylsilane, silanediamine, hexamethyl, silanediamine, n,n,n',n',1,1-hexamethyl, acmc-1ctl2, bis-dimethylamino dimethylsilane, n,n,n,n-hexamethylsilanediamine #, dimethylsilane, bis n,n-dimethylamino, n,n,n',n',1,1-hexamethylsilanediamine
PubChem CID 77384
IUPAC Name N-[dimethylamino(dimethyl)silyl]-N-methylmethanamine
SMILES CN(C)[Si](C)(C)N(C)C

Specifications

Color Yellow
Boiling Point 128°C
Quantity 25 mL
UN Number 2924
Formula Weight 146.31
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(dimethylamino)dimethylsilane