Learn More
Bis(cyclopentadienyl)titanium dichloride, 99+%
CAS: 1271-19-8 | C10H10Cl2Ti-2 | 248.957 g/mol
Supplier: Thermo Scientific Chemicals 04038609
Description
This metallocene is a common reagent in organometallic and organic synthesis. Titanocene dichloride is used to prepare titanocene pentasulfide. Used as an anticancer drug.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
| Bis(cyclopentadienyl)titanium dichloride | |
| ∼285°C (decomposition) | |
| 1.60 g/mL | |
| C10H10Cl2Ti-2 | |
| 10 g | |
| 14,9482 | |
| Reacts in moist air and in water to form HCl . Slightly hydrolysis in water. | |
| [CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2] | |
| 248.957 | |
| 249 | |
| Crystalline |
| 1271-19-8 | |
| Red | |
| Odorless | |
| MFCD00003723 | |
| Air and moisture sensitive | |
| Dichlorobis(cyclopentadienyl)titanium; Titanocene dichloride | |
| MKNXBRLZBFVUPV-UHFFFAOYSA-L | |
| cyclopenta-1,3-diene;titanium(2+);dichloride | |
| 124040768 | |
| ≥99% |
Chemical Identifiers
| 1271-19-8 | |
| 248.957 | |
| MKNXBRLZBFVUPV-UHFFFAOYSA-L | |
| 124040768 | |
| [CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2] |
| C10H10Cl2Ti-2 | |
| MFCD00003723 | |
| Dichlorobis(cyclopentadienyl)titanium; Titanocene dichloride | |
| cyclopenta-1,3-diene;titanium(2+);dichloride |
Safety and Handling
GHS H Statement
H341-H302-H332-H315-H319-H335
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 215-035-9
RTECSNumber : XR2050000
TSCA : Yes
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only