missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)titanium dichloride, 99+%

Catalog No. p-7051064
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4038604 2 g
AA4038609 10 g
AA4038618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4038604 Supplier Thermo Scientific Chemicals Supplier No. 04038604

CAS: 1271-19-8 | C10H10Cl2Ti-2 | 248.957 g/mol

This metallocene is a common reagent in organometallic and organic synthesis. Titanocene dichloride is used to prepare titanocene pentasulfide. Used as an anticancer drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon

Chemical Identifiers

CAS 1271-19-8
Molecular Formula C10H10Cl2Ti-2
Molecular Weight (g/mol) 248.957
MDL Number MFCD00003723
InChI Key MKNXBRLZBFVUPV-UHFFFAOYSA-L
Synonym Dichlorobis(cyclopentadienyl)titanium; Titanocene dichloride
PubChem CID 124040768
IUPAC Name cyclopenta-1,3-diene;titanium(2+);dichloride
SMILES [CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2]

Specifications

Melting Point ∼285°C (decomposition)
Color Red
Density 1.60 g/mL
Odor Odorless
Quantity 2 g
Sensitivity Air and moisture sensitive
Merck Index 14,9482
Solubility Information Reacts in moist air and in water to form HCl . Slightly hydrolysis in water.
Formula Weight 249
Percent Purity ≥99%
Physical Form Crystalline
Chemical Name or Material Bis(cyclopentadienyl)titanium dichloride
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H341-H302-H332-H315-H319-H335
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 215-035-9
RTECSNumber XR2050000
TSCA Yes
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only