missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)dimethyltitanium, 5 wt.% in toluene, AcroSeal™

Catalog No. AC301761000
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog No. Quantity Packaging
AC301761000 100 mL AcroSeal™ Glass Bottle
1 options

Catalog No. AC301761000

Supplier: Thermo Scientific Chemicals 301761000

Only null left

CAS: 1271-66-5 | C12H20Ti | 212.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 1271-66-5 , 108-88-3
Molecular Formula C12H20Ti
Molecular Weight (g/mol) 212.159
MDL Number MFCD02093882
InChI Key CRKLPNOLYBORCP-UHFFFAOYSA-N
Synonym bis cyclopentadienyl dimethyltitanium, dimethyltitanocene in tetrahydrofuran/toluene
PubChem CID 131674729
IUPAC Name cyclopenta-1,3-diene;methane;titanium
SMILES C.C.C1C=CC=C1.C1C=CC=C1.[Ti]

Specifications

Color Orange to Red
Density 0.9g/mL
Packaging AcroSeal™ Glass Bottle
Quantity 100 mL
Formula Weight 208.14
Physical Form Solution
Chemical Name or Material Bis(cyclopentadienyl)dimethyltitanium, 5 wt. % solution in Toluene
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause drowsiness or dizziness.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Suspected of damaging the unborn child.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only