missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclohexanone) Oxalyldihydrazone 98.0+%, TCI America™

Catalog No. B04761G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B04761G 1 g
B047625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B04761G Supplier TCI America Supplier No. B04761G

Chemical Identifiers

CAS 370-81-0
Molecular Formula C14H22N4O2
Molecular Weight (g/mol) 278.356
MDL Number MFCD00001659
InChI Key DSRJIHMZAQEUJV-UHFFFAOYSA-N
Synonym cuprizone, cuprizane, biscyclohexanone oxaldihydrazone, ethanedioic acid, bis cyclohexylidenehydrazide, oxalic acid bis cyclohexylidenehydrazide, biscyclohexanone oxalyldihydrazone, bis cyclohexanone oxaldihydrazone, cuprizon, unii-5n16u7e0ao, oxalic acid, bis cyclohexylidenehydrazide
PubChem CID 9723
IUPAC Name N,N'-bis(cyclohexylideneamino)oxamide
SMILES C1CCC(=NNC(=O)C(=O)NN=C2CCCCC2)CC1

Specifications

Melting Point 208°C
Color White-Yellow
Quantity 1 g
Formula Weight 278.36
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(cyclohexanone) Oxalyldihydrazone
RTECSNumber RO2520000
TSCA Yes