missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclohexanone) oxaldihydrazone, 98%

Catalog No. AAA1062814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1062814 25 g
AAA1062822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1062814 Supplier Thermo Scientific Chemicals Supplier No. A1062814

CAS: 370-81-0 | C14H22N4O2 | 278.356 g/mol

Bis(cyclohexanone) oxaldihydrazone is used in the quantitative spectroscopic determination of copper. It is also essential for the experimental study of mitochondrial metabolism. It is used to study the human demyelinating diseases such as multiple sclerosis and components of neuroinflammation. Further, it acts as ligand and catalyzes Ullmann-type C-N bond forming reactions in the presence of copper salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(cyclohexanone) oxaldihydrazone is used in the quantitative spectroscopic determination of copper. It is also essential for the experimental study of mitochondrial metabolism. It is used to study the human demyelinating diseases such as multiple sclerosis and components of neuroinflammation. Further, it acts as ligand and catalyzes Ullmann-type C-N bond forming reactions in the presence of copper salt.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 370-81-0
Molecular Formula C14H22N4O2
Molecular Weight (g/mol) 278.356
MDL Number MFCD00001659
InChI Key DSRJIHMZAQEUJV-UHFFFAOYSA-N
Synonym cuprizone, cuprizane, biscyclohexanone oxaldihydrazone, ethanedioic acid, bis cyclohexylidenehydrazide, oxalic acid bis cyclohexylidenehydrazide, biscyclohexanone oxalyldihydrazone, bis cyclohexanone oxaldihydrazone, cuprizon, unii-5n16u7e0ao, oxalic acid, bis cyclohexylidenehydrazide
PubChem CID 9723
IUPAC Name N,N'-bis(cyclohexylideneamino)oxamide
SMILES C1CCC(=NNC(=O)C(=O)NN=C2CCCCC2)CC1

Specifications

Melting Point 208°C to 212°C
Quantity 25 g
Beilstein 2388004
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 278.36
Percent Purity 98%
Chemical Name or Material Bis (cyclohexanone) oxaldihydrazone
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 206-729-2
RTECSNumber RO2520000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only