missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(catecholato)diboron 98.0+%, TCI America™
$78.29 - $268.43
Chemical Identifiers
| CAS | 13826-27-2 |
|---|---|
| Molecular Formula | C12H8B2O4 |
| Molecular Weight (g/mol) | 237.812 |
| InChI Key | WYBQOWXCLDXZNR-UHFFFAOYSA-N |
| PubChem CID | 3488065 |
| IUPAC Name | 2-(1,3,2-benzodioxaborol-2-yl)-1,3,2-benzodioxaborole |
| SMILES | B1(OC2=CC=CC=C2O1)B3OC4=CC=CC=C4O3 |
Chemical Identifiers
| 13826-27-2 | |
| 237.812 | |
| 3488065 | |
| B1(OC2=CC=CC=C2O1)B3OC4=CC=CC=C4O3 |
| C12H8B2O4 | |
| WYBQOWXCLDXZNR-UHFFFAOYSA-N | |
| 2-(1,3,2-benzodioxaborol-2-yl)-1,3,2-benzodioxaborole |
Specifications
| 13826-27-2 | |
| White-Yellow | |
| 1 g | |
| WYBQOWXCLDXZNR-UHFFFAOYSA-N | |
| 2-(1,3,2-benzodioxaborol-2-yl)-1,3,2-benzodioxaborole | |
| 3488065 | |
| ≥98.0% (GC) | |
| Bis(catecholato)diboron |
| 196°C | |
| C12H8B2O4 | |
| 2,2′C-Bi-1,3,2-benzodioxaborole | |
| B1(OC2=CC=CC=C2O1)B3OC4=CC=CC=C4O3 | |
| 237.812 | |
| 237.81 | |
| Crystalline Powder |
Safety and Handling
TSCA : No