accessibility menu, dialog, popup

UserName

BOOBOO

CAS RN 13826-27-2

IUPAC Name: 2-(2H-1,3,2-benzodioxaborol-2-yl)-2H-1,3,2-benzodioxaborole
Synonyms: 2-(1,3,2-benzodioxaborol-2-yl)-1,3,2-benzodioxaborole
Molecular Weight (g/mol): 237.81
Molecular Formula: C12H8B2O4
InChi Key: WYBQOWXCLDXZNR-UHFFFAOYSA-N
SMILES: O1B(OC2=CC=CC=C12)B1OC2=CC=CC=C2O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results