missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(catecholato)diboron 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 196°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Synonym | 2,2′C-Bi-1,3,2-benzodioxaborole |
| Formula Weight | 237.81 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Bis(catecholato)diboron |
Safety and Handling
| TSCA | No |