missing translation for 'onlineSavingsMsg'
Learn More

Bis(carboxyethylgermanium) sesquioxide, 99.7%

CAS: 12758-40-6 | C6H10Ge2O7 | 339.399 g/mol

$136.97 - $1445.78

Chemical Identifiers

CAS 12758-40-6
Molecular Formula C6H10Ge2O7
Molecular Weight (g/mol) 339.399
MDL Number MFCD00049319
InChI Key XEABSBMNTNXEJM-UHFFFAOYSA-N
Synonym propagermanium, proxigermanium, carboxyethylgermanium sesquioxide, 14c-proxigermanium, 2-carboxyethylgermasesquioxane, dipropanoic acid germanium sequioxide, 2-carboxyethyl germanium sesquioxide, bis 2-carboxyethylgermanium sesquioxide, ccris 5469, carboxylethylgermanium sesquioxide
PubChem CID 83030
ChEBI CHEBI:32060
IUPAC Name 3-[[2-carboxyethyl(oxo)germyl]oxy-oxogermyl]propanoic acid
SMILES C(C[Ge](=O)O[Ge](=O)CCC(=O)O)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3675806
View Documents
Thermo Scientific Chemicals
03675806
5 g
Each for $136.97
Only null left
 
AA3675814
View Documents
Thermo Scientific Chemicals
03675814
25 g
Each for $451.92
Only null left
 
AA3675822
View Documents
Thermo Scientific Chemicals
03675822
100 g
Each for $1,445.78
Only null left
 
Description

Description

A bifunctional metalloligand with metal-C (Ge-C) bonds,bis(carboxyethylgermanium) sesquioxide, has been used to make two metal-organometallic frameworks (MOMFs). bis-(carboxyethylgermanium)sesquioxide is a as a promising anti-oxidative agent. Bis(carboxyethylgermanium) sesquioxide induce INF-γ and has undergone clinical trials with promising preclinical results.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12758-40-6
339.399
XEABSBMNTNXEJM-UHFFFAOYSA-N
83030
3-[[2-carboxyethyl(oxo)germyl]oxy-oxogermyl]propanoic acid
C6H10Ge2O7
MFCD00049319
propagermanium, proxigermanium, carboxyethylgermanium sesquioxide, 14c-proxigermanium, 2-carboxyethylgermasesquioxane, dipropanoic acid germanium sequioxide, 2-carboxyethyl germanium sesquioxide, bis 2-carboxyethylgermanium sesquioxide, ccris 5469, carboxylethylgermanium sesquioxide
CHEBI:32060
C(C[Ge](=O)O[Ge](=O)CCC(=O)O)C(=O)O

Specifications

12758-40-6
Odorless
MFCD00049319
propagermanium, proxigermanium, carboxyethylgermanium sesquioxide, 14c-proxigermanium, 2-carboxyethylgermasesquioxane, dipropanoic acid germanium sequioxide, 2-carboxyethyl germanium sesquioxide, bis 2-carboxyethylgermanium sesquioxide, ccris 5469, carboxylethylgermanium sesquioxide
XEABSBMNTNXEJM-UHFFFAOYSA-N
3-[[2-carboxyethyl(oxo)germyl]oxy-oxogermyl]propanoic acid
83030
339.25
Powder
320°C (decomposition)
C6H10Ge2O7
5 g
Slightly soluble in water.
C(C[Ge](=O)O[Ge](=O)CCC(=O)O)C(=O)O
339.399
CHEBI:32060
99.7%
Bis(carboxyethylgermanium) sesquioxide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 235-800-0

RTECSNumber : UA2459750

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only