accessibility menu, dialog, popup

UserName

CAS RN 12758-40-6

IUPAC Name: 3-({[(2-carboxyethyl)(oxo)germyl]oxy}(oxo)germyl)propanoic acid
Synonyms: propagermanium carboxylethylgermanium sesquioxide carboxyethylgermanium sesquioxide dicarboxyethenylgermanium sesquioxide 2-carboxyethylgermasesquioxane 丙帕锗 dipropanoic acid germanium sequioxide repagermanium dcefsqd proxigermanium
Molecular Weight (g/mol): 339.4
Molecular Formula: C6H10Ge2O7
InChi Key: XEABSBMNTNXEJM-UHFFFAOYSA-N
SMILES: OC(=O)CC[Ge](=O)O[Ge](=O)CCC(O)=O
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)