missing translation for 'onlineSavingsMsg'
Learn More

Bis(benzonitrile)dichloroplatinum(II), Pt 40% min

Catalog No. p-7048275
Change view
Click to view available options
Quantity:
0.5 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1000402 0.5 g
AA1000403 1 g
AA1000406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1000402 Supplier Thermo Scientific Chemicals Supplier No. 01000402

CAS: 14873-63-3 | C14H10Cl2N2Pt | 472.23 g/mol

Bis(benzonitrile)dichloroplatinum(II) is used as a catalyst for asymmetric hydroformylation reactions, allylation reactions, cyclopropanation reactions, hydrosilylation reactions and carbene insertion into O-H bonds of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14873-63-3
Molecular Formula C14H10Cl2N2Pt
Molecular Weight (g/mol) 472.23
MDL Number MFCD00013125
InChI Key FMBJZWFXNZMJPC-UHFFFAOYSA-L
Synonym dichlorobis benzonitrile platinum ii, bis benzonitrile dichloroplatinum, bis benzonitrile dichloroplatinum ii, cis-bis benzonitrile dichloroplatinum ii, benzonitrile; dichloroplatinum, ptcl2 phcn 2, dichlorobis benzonitrile platinum, dichloro-bis-benzonitrile platinum ii, dichlorobis benzonitrile ;platinum ii ;, bis benzonitrile dichloroplatinate ii
PubChem CID 6093685
IUPAC Name benzonitrile;dichloroplatinum
SMILES Cl[Pt++]Cl.N#CC1=CC=CC=C1.N#CC1=CC=CC=C1

Specifications

Quantity 0.5 g
UN Number UN3439
Solubility Information Slightly soluble in acetone, chloroform
Formula Weight 472.25
Concentration or Composition (by Analyte or Components) Pt 40% min.
Physical Form Powder
Chemical Name or Material Bis(benzonitrile)dichloroplatinum(II)
Hazard Category H301+H311-H317-H330-H334
Hazard Statement GHS H Statement
H301-H311-H331-H334-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P284-P285-P301+P310-P302+P352-P304+P340-P310-P312-P330-P333+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 238-943-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only