accessibility menu, dialog, popup

UserName

NNPtClCl

CAS RN 14873-63-3

IUPAC Name: bis(benzonitrile); dichloroplatinumbis(ylium)
Synonyms: bis(benzonitrile); dichloroplatinumbis(ylium)
Molecular Weight (g/mol): 472.23
Molecular Formula: C14H10Cl2N2Pt
InChi Key: FMBJZWFXNZMJPC-UHFFFAOYSA-L
SMILES: Cl[Pt++]Cl.N#CC1=CC=CC=C1.N#CC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)