missing translation for 'onlineSavingsMsg'
Learn More

Bis(acetonitrile)palladium(II) Dichloride 98.0+%, TCI America™

Catalog No. B16761G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16761G 1 g
B16765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16761G Supplier TCI America Supplier No. B16761G

Chemical Identifiers

CAS 14592-56-4
Molecular Formula C4H6Cl2N2Pd
Molecular Weight (g/mol) 259.42
MDL Number MFCD00013122
InChI Key XGPJSRXOBCGBMN-UHFFFAOYSA-L
Synonym Dichlorobis(acetonitrile)palladium(II)
IUPAC Name bis(acetonitrile); dichloropalladiumbis(ylium)
SMILES CC#N.CC#N.Cl[Pd++]Cl

Specifications

Quantity 1 g
UN Number 3439
Formula Weight 259.43
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(acetonitrile)palladium(II) Dichloride