missing translation for 'onlineSavingsMsg'
Learn More

Bis(7-methyloctyl)Â phthalate, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013487
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013487 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013487 Supplier Merck Emd Millipore Supplier No. 4956850MG
Environmental

Specifications

CAS 20548-62-3
Linear Formula C26H42O4
MDL Number MFCD00044119
Synonym DINP; Diisononyl phthalate; Phthalic acid bis(7-methyloctyl) ester
Type Analytical Standard
Quantity 50 mg
Formula Weight 418.61
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard