missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(4-nitrophenyl) Phosphate 98.0+%, TCI America™
$573.70 - $3193.85
Chemical Identifiers
| CAS | 645-15-8 |
|---|---|
| Molecular Formula | C12H9N2O8P |
| Molecular Weight (g/mol) | 340.18 |
| MDL Number | MFCD00007318 |
| InChI Key | MHSVUSZEHNVFKW-UHFFFAOYSA-N |
| Synonym | Phosphoric Acid Bis(4-nitrophenyl) Ester |
| PubChem CID | 255 |
| ChEBI | CHEBI:3122 |
| IUPAC Name | bis(4-nitrophenoxy)phosphinic acid |
| SMILES | OP(=O)(OC1=CC=C(C=C1)[N+]([O-])=O)OC1=CC=C(C=C1)[N+]([O-])=O |
Chemical Identifiers
| 645-15-8 | |
| 340.18 | |
| MHSVUSZEHNVFKW-UHFFFAOYSA-N | |
| 255 | |
| bis(4-nitrophenoxy)phosphinic acid |
| C12H9N2O8P | |
| MFCD00007318 | |
| Phosphoric Acid Bis(4-nitrophenyl) Ester | |
| CHEBI:3122 | |
| OP(=O)(OC1=CC=C(C=C1)[N+]([O-])=O)OC1=CC=C(C=C1)[N+]([O-])=O |
Specifications
| 645-15-8 | |
| White | |
| MFCD00007318 | |
| Phosphoric Acid Bis(4-nitrophenyl) Ester | |
| OP(=O)(OC1=CC=C(C=C1)[N+]([O-])=O)OC1=CC=C(C=C1)[N+]([O-])=O | |
| 340.18 | |
| CHEBI:3122 | |
| ≥98.0% (T) | |
| Bis(4-nitrophenyl) Phosphate [for Phosphodiesterase Substrate] |
| 176°C | |
| C12H9N2O8P | |
| 1 g | |
| MHSVUSZEHNVFKW-UHFFFAOYSA-N | |
| bis(4-nitrophenoxy)phosphinic acid | |
| 255 | |
| 340.18 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : TB8260000
TSCA : Yes