accessibility menu, dialog, popup

UserName

CAS RN 645-15-8

OHPOONOOONOO

CAS RN 645-15-8

IUPAC Name: bis(4-nitrophenoxy)phosphinic acid
Synonyms: bis(p-nitrophenyl)phosphate
Molecular Weight (g/mol): 340.18
Molecular Formula: C12H9N2O8P
InChi Key: MHSVUSZEHNVFKW-UHFFFAOYSA-N
SMILES: OP(=O)(OC1=CC=C(C=C1)[N+]([O-])=O)OC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results