missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-hydroxyphenyl) Sulfone 98.0+%, TCI America™

Catalog No. p-7118481
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B049525G 25 g
B0495500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B049525G Supplier TCI America Supplier No. B049525G
Only null left

Chemical Identifiers

CAS 80-09-1
Molecular Formula C12H10O4S
Molecular Weight (g/mol) 250.27
MDL Number MFCD00002350
InChI Key VPWNQTHUCYMVMZ-UHFFFAOYSA-N
Synonym 4,4'-sulfonyldiphenol, bisphenol s, bis 4-hydroxyphenyl sulfone, phenol, 4,4'-sulfonylbis, 4,4'-dihydroxydiphenyl sulfone, 4-hydroxyphenyl sulfone, bis p-hydroxyphenyl sulfone, 4,4'-sulfonylbisphenol, diphone c
PubChem CID 6626
ChEBI CHEBI:34372
IUPAC Name 4-(4-hydroxybenzenesulfonyl)phenol
SMILES OC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 247°C
Color White
Quantity 25 g
Formula Weight 250.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-hydroxyphenyl) Sulfone
EINECSNumber (3)-2169
RTECSNumber SM8925000
TSCA Yes