missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-hydroxyphenyl) Sulfide 98.0+%, TCI America™

Catalog No. D135625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D135625G 25 g
D1356500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D135625G Supplier TCI America Supplier No. D135625G

Chemical Identifiers

CAS 2664-63-3
Molecular Formula C12H10O2S
Molecular Weight (g/mol) 218.27
MDL Number MFCD00002349
InChI Key VWGKEVWFBOUAND-UHFFFAOYSA-N
Synonym 4,4'-thiodiphenol, bis 4-hydroxyphenyl sulfide, thiobisphenol, 4,4'-thiobis-phenol, phenol, 4,4'-thiobis, 4,4'-thiobisphenol, 4,4'-dihydroxydiphenyl sulfide, bis 4-oxyphenyl sulfide, p,p'-dihydroxydiphenyl sulfide, 4,4'-dioxydiphenyl sulfide
PubChem CID 17570
ChEBI CHEBI:38957
IUPAC Name 4-[(4-hydroxyphenyl)sulfanyl]phenol
SMILES OC1=CC=C(SC2=CC=C(O)C=C2)C=C1

Specifications

Melting Point 152°C
Quantity 25 g
Formula Weight 218.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-hydroxyphenyl) Sulfide
EINECSNumber (3)-4030
RTECSNumber SN0800000
TSCA Yes