missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(4-hydroxyphenyl) Disulfide 98.0+%, TCI America™
$65.24 - $165.90
Chemical Identifiers
| CAS | 15015-57-3 |
|---|---|
| Molecular Formula | C12H10O2S2 |
| Molecular Weight (g/mol) | 250.33 |
| MDL Number | MFCD00020164 |
| InChI Key | XGKGITBBMXTKTE-UHFFFAOYSA-N |
| PubChem CID | 280665 |
| IUPAC Name | 4-[(4-hydroxyphenyl)disulfanyl]phenol |
| SMILES | C1=CC(=CC=C1O)SSC2=CC=C(C=C2)O |
Chemical Identifiers
| 15015-57-3 | |
| 250.33 | |
| XGKGITBBMXTKTE-UHFFFAOYSA-N | |
| 4-[(4-hydroxyphenyl)disulfanyl]phenol |
| C12H10O2S2 | |
| MFCD00020164 | |
| 280665 | |
| C1=CC(=CC=C1O)SSC2=CC=C(C=C2)O |
Specifications
| 15015-57-3 | |
| Yellow | |
| MFCD00020164 | |
| 4,4′C-Dihydroxydiphenyl Disulfide, 4,4′C-Dithiodiphenol | |
| C1=CC(=CC=C1O)SSC2=CC=C(C=C2)O | |
| 250.33 | |
| 250.33 | |
| Crystalline Powder |
| 152°C | |
| C12H10O2S2 | |
| 1 g | |
| XGKGITBBMXTKTE-UHFFFAOYSA-N | |
| 4-[(4-hydroxyphenyl)disulfanyl]phenol | |
| 280665 | |
| ≥98.0% (GC) | |
| Bis(4-hydroxyphenyl) Disulfide |
Safety and Handling
TSCA : No