missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-formylphenyl)phenylamine (purified by sublimation) 95.0+%, TCI America™

Catalog No. B28431G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B28431G Supplier TCI America Supplier No. B28431G
Only null left

Chemical Identifiers

CAS 53566-95-3
Molecular Formula C20H15NO2
Molecular Weight (g/mol) 301.35
MDL Number MFCD03070375
InChI Key DOUAFMIJGIUWJX-UHFFFAOYSA-N
Synonym N,N-Bis(4-formylphenyl)aniline, 4,4′C-Diformyltriphenylamine, 4,4′C-(Phenylimino)dibenzaldehyde
PubChem CID 819489
IUPAC Name 4-[(4-formylphenyl)(phenyl)amino]benzaldehyde
SMILES O=CC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=O)C=C1

Specifications

Melting Point 142°C
Quantity 1 g
Formula Weight 301.35
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-formylphenyl)phenylamine (purified by sublimation)